Anticorps polyclonal de lapin anti-récepteur du PDGF Alpha
ABCAAB5452-50
: Abcam
- Type d'anticorps:Primaire
- Nom de l'antigène:PDGF Receptor alpha
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:
- Réactivité:
- Western blot:Yes
- Environmentally Preferable:
- Température de stockage:Cool
- Température de transport:Ice
- Cdt:50 µl
:
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type : Primaire
Antigène : Récepteur alpha PDGF
Clonalité : Polyclonal
Clone:
Conjugaison : Non conjugué
Epitope :
Hôte : Lapin
Isotype: IgG
Réactivité : Souris