Anti-KAT13A / SRC1 Rabbit Polyclonal Antibody
Product Details & Documents
Rabbit Polyclonal KAT13A / SRC1 antibody. Suitable for ICC, ICC/IF, IHC-Fr and reacts with Human, Mouse, Rat samples. Cited in 9 publications. Immunogen corresponding to Synthetic Peptide within Human NCOA1 aa 1-50.
Attention: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: KAT13A / SRC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat; Mouse
Spécifications
- Seq:1286
- Taille:100 µg
- Type de conditionnement:Vial
- Type d'anticorps:Primaire
- Formulaire:Liquid
- Purification:Immunogen Affinity purification
- Symbole de l'antigène:KAT13A / SRC1
- Synonymes antigène:SRC1|NCOA1|Nuclear receptor coactivator 1|NCoA-1|Class E basic helix-loop-helix protein 74|Protein Hin-2|RIP160|Renal carcinoma antigen NY-REN-52|Steroid receptor coactivator 1|SRC-1
- ImmunoFluorescence:Yes
- Isotype:IgG
- Masse moléculaire:156757 Da
- Conjugaison:Unconjugated
- Concentration:1 mg/ml
- Réactivité:Human|Rat|Mouse
- Température de stockage:Shipped at 4 °C. Store at +4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C or –80 °C. Avoid freeze / thaw cycle.
- Cat. No.:ABCAAB2859-100
- Clonalité:Polyclonal
- Immunogène:Synthetic Peptide within Human NCOA1 aa 1-50. The exact immunogen used to generate this antibody is proprietary information.
- Nom de l'antigène:BHLHE74
- Hôte:Rabbit
- ImmunoChimie:Yes
- Tested applications:ICC, ICC/IF, IHC-Fr
- Storage buffer:PBS, BSA
- Température de transport:+4 °C



