Auftragserfassung
Switzerland
ContactUsLinkComponent
 
Anticorps monoclonal de souris anti-IgG3 [clone : 5G12]
  ABCAAB1928-500
 :  Abcam
Anticorps monoclonal de souris anti-IgG3 [clone : 5G12]
  ABCAAB1928-500
 :  Abcam

 

  • Type d'anticorps:
    Primaire
  • Symbole de l'antigène:
    IgG3
  • Clonalité:
    Monoclonal
  • Clone:
    5G12
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Mouse
  • Isotype:
  • Réactivité:
  • Environmentally Preferable:
  • Température de stockage:
    Cool
  • Température de transport:
    Ice
  • Cdt:
    500 µG

 

 

 :

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type : Primaire
Antigène : IgG3
clonalité : Monoclonal
Clone: 5G12
Conjugaison: Non conjugué
Epitope :
Hôte : Souris
Isotype: IgG1
Réactivité: Humain