Anti-NFKB2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
BIRBORB9348-100
: Biorbyt
- Type d'anticorps:Primaire
- Nom de l'antigène:Nuclear factor of kappa light polypeptide gene enhancer in B cells 2 (p49/p100)
- Symbole de l'antigène:NFKB2
- Clonalité:Polyclonal
- Conjugaison:FITC (Fluorescein Isothiocyanate)
- Hôte:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Formulaire:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:50/100 kDa
- Température de stockage:Cool
- Concentration:0,5 mg/ml
- Température de transport:Ice
- Immunogène:KLH conjugated synthetic peptide derived from human NFKB p52.
- Purification:purified by affinity chromatography
- Taille:100 µg
- Cdt:100 µG
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: NFKB2 (nuclear factor of kappa light polypeptide gene enhancer in B-cells 2 (p49/p100))
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: