Anti-CD83 Mouse Polyclonal Antibody
50 µl
Catalog # ABNOH00009308-A01
Supplier: Abnova
Jump to:
Product Details & Documents
Type: Primary
Antigen: CD83
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Documents
TDS_ABNOH00009308-A01.pdf
Datasheet
Specifications
- Taille:50 µl
- Western blot:Yes
- Adsorption croisée:No
- Type d'anticorps:Primaire
- Symbole de l'antigène:CD83
- Synonymes antigène:Cell-Surface Glycoprotein|B-Cell Activation Protein|BL11|HB15
- Conjugaison:Unconjugated
- Réactivité:Human
- Température de stockage:Cool
- ID de gène:9308
- Cat. No.:ABNOH00009308-A01
- Clonalité:Polyclonal
- Nom de l'antigène:CD83 molecule
- Hôte:Mouse
- Température de transport:Ice
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



