Anti-LIPM Rabbit Polyclonal Antibody
Catalog # AVIVARP64348_P050
Supplier: Aviva Systems Biology
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:lipase, family member M
- Symbole de l'antigène:LIPM
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Réactivité:Pig,Human
- Western blot:Yes
- Épitope:N-terminal
- Adsorption croisée:none
- ID de gène:340654
- Synonymes antigène:bA304I5.1|LIPL3
- Température de stockage:Cool
- Température de transport:Ice
- Cdt:50 µG
Specifications
About this item
Type: Primary
Antigen: LIPM (lipase, family member M)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Pig