Anti-NR2C1 Rabbit Polyclonal Antibody
Catalog # AVIVARP37896_P050
Supplier: Aviva Systems Biology
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:nuclear receptor subfamily 2, group C, member 1
- Symbole de l'antigène:NR2C1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Western blot:Yes
- Épitope:middle
- Adsorption croisée:none
- ID de gène:22025
- Synonymes antigène:4831444H07Rik|Tr2-11|Eenr|TR2|80.3
- Température de stockage:Cool
- Température de transport:Ice
- Cdt:50 µG
Specifications
About this item
Type: Primary
Antigen: NR2C1 (nuclear receptor subfamily 2, group C, member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: