Anti-HSP90AA1 Rabbit Polyclonal Antibody
Catalog # BOSSBS-3181R
Supplier: Bioss
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Heat shock protein 90kDa alpha (cytosolic), class A member 1
- Symbole de l'antigène:HSP90AA1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Adsorption croisée:none
- Synonymes antigène:HSPCA|Hsp89|EL52|HSP86|LAP-2|HSPCAL1|HSPN|Hsp90|HSPC1|HSP90N|HSPCAL4|LAP2|HSP89A|HSP90A
- Température de stockage:Cool
- Température de transport:Ice
- Cdt:100 µl
Specifications
About this item
Type: Primary
Antigen: HSP90AA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat