Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:bone morphogenetic protein 4
- Symbole de l'antigène:BMP4
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Mouse
- Western blot:Yes
- Adsorption croisée:none
- ID de gène:652
- Synonymes antigène:BMP2B1|MCOPS6|ZYME|OFC11|BMP2B
- Température de stockage:Store for 1 year at –20 °C from date of shipment
- Concentration:1 mg/ml
- Température de transport:Ice
- Immunogène:Synthetic peptide
- Purification:Protein A column
- Taille:100 µl
- Cdt:0,1 mL
Specifications
About this item
Bone morphogenetic protein 4, also known as BMP4, is a member of the bone morphogenetic protein family which is part of the transforming growth factor-β superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo. BMPs also regulate cell proliferation, differentiation, lineage determination, motility, and death.
BMP4 is a potent bone-inducing morphogen, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva.
BMP4 is a critical signaling molecule required for the early differentiation of the embryo and establishing of a dorsal-ventral axis. It also plays a role in the epithelial-mesenchymal interactions leading to tooth formation.
Both BMP2 and BMP4 have been found in calcified atherosclerotic plaques and aortic valve diseases, which suggests their importance in cardiovascular diseases.
Western Blot: 1:2,000;
Type: Primary
Antigen: BMP4 (bone morphogenetic protein 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse