Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Rho-associated, coiled-coil containing protein kinase 1
- Symbole de l'antigène:ROCK1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Adsorption croisée:none
- ID de gène:6093
- Synonymes antigène:ROCK-I|P160ROCK
- Température de stockage:Store for 1 year at –20 °C from date of shipment
- Température de transport:Ice
- Immunogène:Synthetic peptide
- Taille:100 µl
- Cdt:0,1 mL
Spécifications
À propos de cet article
ROCK, Rho-associated coiled-coil forming kinases, is a protein serine/threonine kinase that is activated when bound to the GTP-bound form of Rho. The small GTPase Rho regulates formation of focal adhesions and stress fibers of fibroblasts, as well as adhesion and aggregation of platelets and lymphocytes by shuttling between the inactive GDP-bound form and the active GTP-bound form. Rho is also essential in cytokinesis and plays a role in transcriptional activation by serum response factor. ROCK, as a downstream effector of Rho, phosphorylates and activates LIM kinase, which in turn, phosphorylates cofilin, inhibiting its actin-depolymerizing activity. ROCK1 is involved in diverse cellular functions, including smooth muscle contraction, actin cytoskeleton organization, cell adhesion and motility, and gene expression.
Western Blot: 1:2,000;
Type: Primary
Antigen: ROCK1 (Rho-associated, coiled-coil containing protein kinase 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat