Anti-MSN Mouse Monoclonal Antibody [clone: 17H2]
ABFRLF-MA0367
- Type d'anticorps:Primaire
- Nom de l'antigène:moesin
- Symbole de l'antigène:MSN
- Clonalité:Monoclonal
- Clone:17H2
- Conjugaison:Unconjugated
- Hôte:Mouse
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Adsorption croisée:none
- ID de gène:4478
- Synonymes antigène:HEL70
- Température de stockage:Store for 1 year at –20 °C from date of shipment
- Concentration:1 mg/ml
- Température de transport:Ice
- Immunogène:Recombinant human protein purified from E.coli
- Purification:Protein G purified
- Taille:100 µl
- Cdt:0,1 mL
Moesin (for membrane-organizing extension spike protein) is a member of the EMR protein family which includes ezrin and radixin. ERM proteins appear to function as cross-linkers between plasma membranes and actin-based cytoskeletons.
Western Blot: 0.5 ?g/ml;
Type: Primary
Antigen: MSN (moesin)
Clonality: Monoclonal
Clone: 17H2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human, Mouse, Rat