Order Entry
Switzerland
ContactUsLinkComponent
Anti-NCK1 Mouse Monoclonal Antibody [clone: AF6D7]
Anti-NCK1 Mouse Monoclonal Antibody [clone: AF6D7]
Catalog # ABFRLF-MA0330
Supplier:  AbFrontier
Anti-NCK1 Mouse Monoclonal Antibody [clone: AF6D7]
Catalog # ABFRLF-MA0330
Supplier:  AbFrontier

Specifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    NCK adaptor protein 1
  • Symbole de l'antigène:
    NCK1
  • Clonalité:
    Monoclonal
  • Clone:
    AF6D7
  • Conjugaison:
    Unconjugated
  • Hôte:
    Mouse
  • Précipitation immunologique:
    Yes
  • Isotype:
    IgG2a
  • Réactivité:
    Human,
    Rat
  • Western blot:
    Yes
  • Adsorption croisée:
    none
  • ID de gène:
    4690
  • Synonymes antigène:
    NCKalpha|NCK|nck-1
  • Température de stockage:
    Store for 1 year at –20 °C from date of shipment
  • Température de transport:
    Ice
  • Immunogène:
    Recombinant human protein purified from E.coli (His-NCK2)
  • Purification:
    Ammonium sulfate precipitation
  • Taille:
    100 µl
  • Cdt:
    0,1 mL

Specifications

About this item

NCK1 is one of the adaptor proteins which mediate specific protein-protein interactions in signaling processes. Adaptor proteins usually contain several domains like SH2 (Src homology 2) and SH3 which allow specific interactions with other specific proteins.
NCK1 and NCK2 showing high sequence identity (68%) have three SH3 domains and a C-terminal SH2 domain. Both of them bind receptor tyrosine kinases such as PDGFR and other tyrosine phosphorylated
proteins via their SH2 domains. Various molecules which interact with SH domains of Nck and regulate signaling process of actin cytoskeleton reorganization have been identified. Ncks are thought to have important functions in the development of mesodermal structures during embryogenesis, linked to a role in cell movement and cytoskeletal reorganization.
Nck also have a function in modulating mRNA translation at the level of initiation by interacting eukayotic initiation factor 2 (eIF2). Under the stressed conditions, protein synthesis is reduced by inhibiting the activity of eIF2 through the phosphorylation, transiently inhibiting recycling of eIF2
into its active form.

Western Blot: 1:1,000; Immunoprecipitation: 0.5 ?l;

Type: Primary
Antigen: NCK1 (NCK adaptor protein 1)
Clonality: Monoclonal
Clone: AF6D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Gamma
Reactivity: Human, Rat