Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Hes family bHLH transcription factor 1
- Symbole de l'antigène:HES1
- Clonalité:Monoclonal
- Clone:7H11
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Mouse
- Précipitation immunologique:Yes
- Isotype:IgG2b
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Adsorption croisée:none
- ID de gène:3280
- Synonymes antigène:HHL|HES-1|HRY|bHLHb39
- Température de stockage:Store for 1 year at –20 °C from date of shipment
- Température de transport:Ice
- Immunogène:His-tagged recombinant human Hes1 protein purified from E.coli
- Purification:Ammonium sulfate precipitation
- Taille:100 µl
- Cdt:0,1 mL
Spécifications
À propos de cet article
The Notch signaling pathway is important for cell-cell communication, which involve gene regulation mechanisms that control multiple cell differentiation processes during embryonic and adult life. It critically influences cell proliferation, differentiation, and apoptosis in metazoans. The hairy and enhancer of split (HES) family is a basic helix-loop-helix (bHLH) type trancriptional repressor and acts as Notch effectors by negatively regulating expression of downstream target genes such as tissue-specific transcription factors. HES-1 is an upstream negative regulator of REST expression. Silencing of the transcriptional repressor REST is required for terminal differentiation of neuronal and β-cells.
Recent integrative genomic analyses on HES/HEY family suggest that HES1 and HES3 are target genes of the embryonic stem cell-specific network of transcription factors, and that HES1, HES5, HEY1, HEY2 and HEYL are target genes of Notch signaling pathway.
Western Blot: 1:500~1,000; Immunoprecipitation: 1 ?l;
Type: Primary
Antigen: HES1 (hes family bHLH transcription factor 1)
Clonality: Monoclonal
Clone: 7H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human, Mouse, Rat