Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Myeloperoxidase
- Symbole de l'antigène:MPO
- Clonalité:Monoclonal
- Clone:1A1
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Mouse
- Précipitation immunologique:Yes
- Isotype:IgG1
- Réactivité:Human
- Adsorption croisée:none
- ID de gène:4353
- Température de stockage:Store for 1 year at –20 °C from date of shipment
- Température de transport:Ice
- Immunogène:Purified from human sputum
- Purification:Ammonium sulfate precipitation
- Taille:100 µl
- Cdt:0,1 mL
Spécifications
À propos de cet article
Myeloperoxidase is a major neutrophil protein and is also present in monocytes. In neutrophils, it is stored in azurophilic granules and released during phagocytosis. It is a heme enzyme that uses the superoxide and hydrogen peroxide generated by the neutrophil oxidative burst to produce hypochlorous acid and other reactive oxidants.The produced hypochlorous acid reacts with and destroys bacteria. In many inflammatory pathologies, such as cystic fibrosis and rheumatoid arthritis, neutrophils are also causing tissue damage. MPO is thought to be the most promising cardiac marker at the moment. In addition to that MPO is a good inflammatory biomarker for autoimmune, inflammatory diseases and cancer.
Immunoprecipitation: 2 ?l;
Type: Primary
Antigen: MPO (myeloperoxidase)
Clonality: Monoclonal
Clone: 1A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human