Peqlab, PCR Plates, 384-Well, Skirted
Catalog # 732-3237
Supplier: VWR Peqlab
Specifications
- Nb de puits:384
- Jupe:Fully skirted
- Couleur des puits:incolore
- Seq:01
- Environmentally Preferable:Low Manufacturing Impact
- Certification environnementale:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Cdt:50
- Volume puits:0,04 ml
Specifications
About this item
PP, autoclavable. These high quality, 384-well PCR plates are suitable for high throughput applications, and are designed to be compatible with robotic platforms and automated liquid handling systems.
- Compatible with most 384-well thermal cycler systems and robots (colour-free plates)
- 40 µl maximum well capacity and 25 µl operating volume
- Cut away corner top right and bottom right for easy orientation
- Manufactured from virgin polypropylene under strict quality control guidelines
- Certified free from RNases, DNases and human DNA