Peqlab, PCR Plates, 96-Well, Semi-Skirted, for Automation
Supplier: VWR Collection
Certificates
About this item
PP, autoclavable. Low profile, semi-skirted plates for use on the ABI Fast Block system or the Roche® LightCycler® 480, offering optimal instrument fit and rapid heat transfer for optimal results on these platforms.
- Ultra-thin for effective heat transfer and minimum condensation
- Alphanumeric annotation for quick sample identification
- Certified free from RNase, DNase and human DNA
- Manufactured from virgin polypropylene under strict quality control guidelines
- Available in white, ideal for use in Real-Time PCR
- Cut away corner top left (colour-free plates) or bottom right (white plates) for easy orientation
Specifications
- Catalog No:
- 732-3236
- 732-3203
- Environmentally Preferable:
- Low Manufacturing Impact
- Certification environnementale:
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)