Peqlab, PCR Plates, 96-Well, Skirted, Low Profile
Catalog # 732-3225
Supplier: VWR Peqlab
Specifications
- Nb de puits:96
- Profil:Bas
- Jupe:Fully skirted
- Couleur des puits:incolore
- Environmentally Preferable:Low Manufacturing Impact
- Certification environnementale:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Cdt:25
Specifications
About this item
PP. Low profile plates significantly reduce the dead space between the film and the sample, which reduces the risk of sample condensation for enhanced reaction reproducibility. Full skirt gives greater stability.
- Ultra-thin, low profile design for effective heat transfer and minimum condensation
- Suitable for use with automated systems
- Eight holes in skirt aid plate positioning and removal from thermal cycler block
- Cut-away corner (H1)
- Certified free from DNase, RNase and human genomic DNA