Peqlab, PCR Plates, 96-Well, Semi-Skirted, qPCR
Catalog # 732-3231
Supplier: VWR Peqlab
Specifications
- Nb de puits:96
- Jupe:Semi-skirted
- Couleur des puits:incolore
- Environmentally Preferable:Low Manufacturing Impact
- Certification environnementale:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Cdt:25
Specifications
About this item
PP. Semi-skirted plate especially designed for Real-Time PCR applications.
- Directly compatible with ABI instrument (7000, 7300, 7500, 7700, 7900 HT and 9500 models)
- Ultra-thin for effective heat transfer and minimum condensation
- Black alphanumeric marking for quick sample identification
- Cut corner (A12)
- Certified free from DNase, RNase and human genomic DNA