Autosampler syringes, Dionex™
DION053916
: DIONEX
- Volume:250 µl
- Désignation:Sample syringe, standard
- Cdt:1
This syringe is designed to pipette liquid from one vial to another or to waste and mixes a vial by repeatedly drawing and expelling the vial contents. It can also be used to deliver a sample to the injection valve.