Magnetic stirring bar restrainer, Spinbar®, SP Bel-Art
Catalog # BELAF377861000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Désignation:Stirring bar restrainer, blue
- Cdt:1
Specifications
About this item
The Spinbar® restrainer fits ergonomically in one hand, and when placed alongside the outer wall of a mixing vessel it retains and secures the stirring bar in position.
- Ensures no direct contact with the stirring magnet
- Retains magnetic stirring bars up to 80 mm long