Pipette stand, rotary, SP Bel-Art
Catalog # BELAF189570000
Supplier: Bel-Art Products, a Part of SP
Specifications
- Ø×h:230×480 mm
- Pour:94 pipettes
- Couleur:bleu/gris
- Cdt:1
Specifications
About this item
Rotary pipette stand made of PP and PVC. Holds up to 94 pipettes and rotates on a central vertical axis for convenient selection.
- Rotating disks are 18 cm (7⅛") in diameter
- Lower disc consists of four concentric troughs with tapered sides and open bottoms which cradle the tips protectively
- Upper rotating disc has a grid pattern concentric with the bottom disc keeping the pipettes vertical
- Liquid draining off the pipette tip is retained on top of the stationary base by the outer rim
- Low center of gravity stabilises the 48 cm high stand on the 23 cm diameter base