Cannula with spray nozzle, Dosys™
Catalog # SOCO370.30500
Supplier: Socorex Isba
Specifications
- Ø×L:3×500 mm
- Désignation:Cannula with spray nozzle, Luer-Lock
- Matière:Acier inoxydable
- Cdt:1
Specifications
About this item
This spray nozzle cannula fits on Luer-Lock self-refilling syringes.
Suitable where spraying and nebulising liquid is required.