Test kit, free cyanide
Catalog # HACH201002
Supplier: Hach
CAS Number:
Specifications
- Cdt:1 KIT
- Désignation:Cyanide (free) test kit, CYN-3
- Plage de mesure:0 - 0,3 mg/l CN-
Specifications
About this item
The test kit is suitable for measuring the effectiveness of cyanide removal from waste effluents of metal-cleaning and electroplating baths and various other chemical processes through alkaline or neutral chlorination procedures. Not suitable for complexed cyanides.
- Simple and fast colour comparison method
- Portable test kit for on site measurement
Delivery: Test kit comprises reagents as powder pillows, comparator, comparator disc, tubes and simple equipment for sample treatment and measurement, all in a sturdy carrying case.