Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Homogénéisateurs
Outils de dispersion
Outils de dispersion pour OV5
Outils de dispersion pour OV5
Outils de dispersion pour OV5
Catalog # VELPA00000037
Supplier:
VELP SCIENTIFIC
Expand image
Outils de dispersion pour OV5
Catalog # VELPA00000037
Supplier:
VELP SCIENTIFIC
Specifications
Désignation:
Outil de dispersion pour milieux solides/liquides
Longueur:
220 mm
Seq:
44
Cdt:
1
Stator/Rotor Ø:
30/23 mm
Type:
VSS3FER3
Specifications
About this item
Product Details
Product Details