Immobilised D-galactose for lectin purification
Catalog # 786-391
Supplier: G-Biosciences
CAS Number:
Specifications
- Cdt:5 mL
- Désignation:Immobilised D-galactose resin
- Seq:01
Specifications
About this item
Immobilised D-Galactose is designed for the rapid purification of lectins, galactosidases and other galactose-binding molecules. The resin is ideal for the purification of agglutinins, lectins, toxins, glactose-binding, N-acetylgalactosaminebinding or carbohydrate binding molecules. Specific applications include the purification of galactosidases, C‐type lectins, enterotoxins and cholera toxin.
The resin consists of 6% beaded agarose that is covalently coupled to thio‐α‐D‐galactose and has a binding capacity of >20 mg jacalin/ml settled resin.