Immobilisation kit, SDC™ (Steroid/Drug/Compound)
Catalog # 786-271
Supplier: G-Biosciences
CAS Number:
Specifications
- Cdt:1 KIT
- Désignation:SDC™ (Steroid/Drug/Compound) Immobilization Kit, for 5 columns
Specifications
About this item
SDC™ (Steroid/Drug/Compound) Immobilisation Kit is designed for the immobilisation of steroids, drugs and chemical compounds that lack primary amines, sulfhydryls, carbonyls and other common coupling groups to a solid-phase agarose support for the use in affinity purification. The kit uses Immobilised DADPA (diaminodipropylamine) resin to bind steroids, drugs and chemicals through their active hydrogens.