Abbe refractometer
Abbe refractometer, Novex, 0 - 95 °Bx, 1,30 - 1,70 nD, 0,5 °Bx; 0,0002 nD
N° de catalogue EUMX98.490
Fournisseur: EUROMEX
Aller à
Product Details & Documents
The Abbe laboratory refractometer is a bench-top instrument for high-precision measurements of an index of refraction.
- More accurate than hand refractometers
- Capable of measuring all kinds of concentrations and identifying several types of substances
It can be connected to a water bath for measuring at a controlled temperature and used for determining the refractive index of glass, plastic films. During operating this type of refractometer, the use of a cold light source is recommended. The instrument is equipped with a built-in thermometer and water connection to control fluid temperatures.
This Abbe refractometer has scales for measurement of sugar concentrations and refractive indices.
The use of a cold light source like the LE.5209 is recommended for this type of refractometer.
: Supplied with flight case, thermometer, test plate and adjusting tools.
Spécifications
- Modèle:Novex
- Plage de mesure:0 - 95 °Bx
- Résolution:0,5 °Bx; 0,0002 nD
- Désignation:Abbe refractometer
- Degré de réfraction:1,30 - 1,70 nD
- Cat. No.:EUMX98.490
Accessories
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix