Handheld spectroscopes
Handheld spectroscopes
Supplier:   A.KRÜSS Optronic
Handheld spectroscopes
Supplier:  A.KRÜSS Optronic
Certificates
Handheld spectroscopes
2 Options Available Below
Product Details & Documents
These handheld spectroscopes are designed for checking the line spectrum of gas discharge lamps as well as individual and comparative observations and spectral examinations.
  • Variable slit and angular dispersion (C-F = 7°)
  • Linear disperson of 60 mm for both models
  • Manual option with grating
Used in education for qualitative analysis and measurement of emission and absorption spectra.
Product Family Options
Product Information
  • Désignation
    Modèle
    Plage de mesure
    Availability
    Price
  • Handheld spectroscope
    1501
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Handheld spectroscope
    1504
    400 − 750 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
Accessories
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • KRUE15081
    Scale illumination
    Log in for availability
    1 * 1 ST - SFr. 164.00
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • KRUE1510
    Stand for handheld spectroscopes
    Log in for availability
    1 * 1 ST - SFr. 173.00