Trays, rectangular
450×320×40 mm
N° de catalogue LAMA1631
Fournisseur: LAMAPLAST
Aller à
Product Details & Documents
PS, white.
- Stable, good break-resistance
- Temperature resistant from −178 to 80 °C (melamine) and −40 to 55 °C (PS)
- Smooth surfaces for easy cleaning
Multipurpose dishes for a wide variety of laboratory applications. Also suitable for use in the food industry.
Spécifications
- l×p×h:450×320×40 mm