Protease inhibitor cocktail, FOCUS™ ProteaseArrest™
Catalog # 733-1945
Supplier: G-Biosciences
CAS Number:
Specifications
- Cdt:1 mL
- Désignation:FOCUS™ ProteaseArrest™ (100X)
Specifications
About this item
A ready to use, 100X concentrated, broad range protease inhibitor cocktail that is fully compatible with 2D electrophoresis and subsequent mass spectrometry.
- Compatible to 2D gel electrophoresis
- Broad spectrum protease inhibitor cocktail
- 100X concentrated, ready to use solution
- Inhibition of protease activity in protein preparations
- Suitable for 2D gel sample preparation for protection of protein samples