0,00 SFr.
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
Pipettes
Pipettes jaugées
Bulb pipettes, one mark, Class B
Bulb pipettes, one mark, Class B
NEUB0272-30-200
:
NEUBERT VOLUME GLASSWAERE
Bulb pipettes, one mark, Class B
NEUB0272-30-200
:
NEUBERT VOLUME GLASSWAERE
Capacité:
200 ml
Cdt:
1
Glass.
Calibrated to deliver 'EX'
Full delivery
Blue graduations
:
DIN 12691