Flasks, round bottom, with three necks and standard ground joints
Catalog # NEUB0759-0025-14
Supplier: NEUBERT VOLUME GLASSWAERE
Specifications
- Capacité:25 ml
- Cdt:1
- Col latéral:14/23 NS
- Col central:14/23 NS
Specifications
About this item
Borosilicate glass 3.3, clear, with angled side necks.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.