Scrubbing brush
Scrubbing brush
Scrubbing brush, without handle, Bois, 220 mm, 50 mm
N° de catalogue NEOL2-3275
Fournisseur:   NEOLAB
Scrubbing brush
N° de catalogue NEOL2-3275
Fournisseur:  NEOLAB
Scrubbing brush
Scrubbing brush
Scrubbing brush, without handle, Bois, 220 mm, 50 mm
Prix
SFr. 5.40
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Scrubbing brush with fibre bristles and wooden block.
Spécifications
  • Handle material:
    Bois
  • Désignation:
    Scrubbing brush, without handle
  • Longueur totale:
    220 mm
  • Longueur brosse:
    50 mm