Contact plate, with grid, ContactClik™
Catalog # 391-3561
Supplier: PHOENIX BIOMEDICAL
Specifications
- Ø:55 mm
- Emballage:20/sleeve, 30 sleeves/case
- Stérilité:Non stérile
- Version:Contact plate (domed base) with grid
- Hauteur:15 mm
- Cdt:600
Specifications
About this item
Used for testing bioburden counts on working surfaces.
- ContactClik™ design allows easy locking and reopening, no orientation required
- Domed surface and media gripping tabs help the media to stick to the dish, reducing drop-off
- Special double ring around the outside of the dish helps prevent fingers from touching the media, thus reducing the possibilities of contamination
- Pre-treated all around the sides of the base and ready for printing a barcode, lot number or any other information as required