ContactUsLinkComponent
SPE plates, 96-well, HyperSep™, Retain-AX
SPE plates, 96-well, HyperSep™, Retain-AX
N° de catalogue 549-0683
SPE plates, 96-well, HyperSep™, Retain-AX
N° de catalogue 549-0683
Spécifications
  • Désignation:
    96-well plate, HyperSep™ Retain-AX
  • Seq:
    06
  • Remplissage:
    60 mg
  • Cdt:
    1
Spécifications
À propos de cet article
HyperSep Retain-AX is a high purity, highly porous polystyrene DVB material partially modified with quaternary functional groups to retain acidic analytes. The problems associated with column drying in traditional silica-based SPE materials are eliminated when using this product.
  • Easy recoveries for acidic analytes
  • High and consistent recoveries
  • High capacity material