Disinfectant, concentrate, Bomix® plus
BODE974602
- Capacité:2 L
- Désignation:Bomix® plus, aldehyde-free disinfectant
- Seq:01
- Emballage:Bottle
- Cdt:2 L
Bomix® plus is used for manual reprocessing of heat-sensitive and heat-resistant instruments in immersion baths and for the coarse and brush cleaning of endoscopes. Also suitable for flexible endoscopes heavily contaminated with protein.
- Contents in 100 g: N,N-Didecyl-N-methyl-poly(oxyethyl)ammoniumpropionate 17,5 g and other ingredients like surfactants, solvents, complexing agents, corrosion inhibitors, foam regulators, pH regulators, colourants and fragrances
- Bactericidal, fungicidal, virucidal against enveloped viruses, polyomavirus and rotavirus
- pH value for 0,3%, 1%, 2% of solution: approx 7
- Suitable for ultrasonic baths