Disinfectants, concentrate, Mikrobac® food
BODL980221
: Bode
- Désignation:Mikrobac® food, aldehyde-free disinfectant for kitchen, food processing and surfaces
- Seq:03
- Emballage:5 l canister
- Cdt:5 l
Mikrobac® food is suitable for the daily disinfecting maintenance cleaning of all water-resistant surfaces in industrial kitchens and food-processing areas including dairy and the dairy processing industry.
- Aldehyde- and fragrance-free
- Good cleaning capacity and material compatibility
- pH value of 1% solution: 8
- Easy to rinse-off
: Bactericide EN 1040: Fungicide EN 1275; Virucidal (DVV) against enveloped viruses (incl. HBV, HIV, HCV)