Extraction unit E-812/E-816
Extraction unit E-812/E-816
 :   BUCHI
Extraction unit E-812/E-816
 :  BUCHI
Extraction unit E-812/E-816
Extraction unit E-812/E-816
Soxhlet extraction for exhaustive total fat extraction with preceding hydrolysis
  • Run six samples simultaneously with E-816 model
  • Reduced solvent consumption and lower running costs
  • High speed heating with ceramic plates and individual heating zones
Product Family Options
  • Désignation
    Type
    Capacité extracteur
    Puissance
  • Extraction Units E-816 SOX
    E-812
    190 ml
    1200 W
    1 * 1 ST
    1 * 1 ST
  • Extraction units E-812 SOX
    E-816
    190 ml
    1200 W
    1 * 1 ST
    1 * 1 ST
Accessories
  •  
    Désignation
  • BUCH049427
    Beaker Soxhlet / ECE, 80 mm, 130 ml
    1 * 2 ST - SFr. 229.90
  •  
    Désignation
  • BUCH018105
    Paper thimbles, 25×100 mm
    1 * 25 ST - SFr. 155.00
  •  
    Désignation
  • BUCH047604
    Condenser
    1 * 1 ST - SFr. 594.70