Autoclaves, vertical, HV
Supplier: HMC Europe GmbH
Certificates
About this item
The four HV models are simple and robost which consists of a quicklock, dual sensor system, themolock and a exhaust air filter and are used for higher standards.
- The important programs are preinstalled and can be done at a push of a button
- Simple and casters for mobile use
- Innovative, passive sealing system
- Manual lid lock - one-handed operation
- Quadruple fail-safe heating system with maximum efficiency
Specifications
- Catalog No:
- 481-4715
- 481-4710
- 481-4705
- 481-4700
- Modèle:
- HV 110
- HV 85
- HV 50
- HV 25
- Capacité:
- 110 L
- 85 L
- 50 L
- 25 L
- Température:
- 127 °C
- 133 °C
- 139 °C
- 130 °C
- Pression max.:
- 1,2 bar
- 1,6 bar
- 2,2 bar
- 1,4 bar
- Ø×h intérieur:
- 420×795 mm
- 420×615 mm
- 300×710 mm
- 240×550 mm
- L×P×H ext.:
- 660×650×1180 mm
- 660×650×1000 mm
- 540×530×1040 mm
- 480×460×950 mm
- Poids:
- 81 kg
- 71 kg
- 57 kg
- 41 kg
- Puissance calorifique:
- 4000 W
- 3000 W
- 2000 W
- 1500 W
- Minuterie:
- +
- +
- +
- +