Face shields, medical Assistant®
Catalog # HECH49075010
Supplier: GLASWARENFABRIK KARL HECHT
Specifications
- Désignation:Face shield for medical applications
- Matière:PC
- Environmentally Preferable:
- Cdt:1
Specifications
About this item
This disposable face shield consists of a headband band made of non allergenic fleece and a transparent PC visor. The face shield covers the entire face, not just the area around the eyes (for example, like safety glasses). This considerably reduces the risk of infection. It is ideal for use in orthopaedics, during general operations, in surgery, urology and dentistry. The shield can also be used by emergency services, as well as in laboratories and emergency wards.
- Can be worn together with safety glasses
- Protective film prevents scratches prior to use
- The moisture-absorbing material of the strap adequately absorbs any perspiration on the brow or temples
- Velcro fastening for a custom fit
- Large field of view
- Prevents direct inhalation of smoke when using a cauter