Buckets with Lid, Round
Fournisseur: SEMADENI
Certificats
À propos de cet article
PP, white.
- Stackable and sturdy with plastic carrying handle
- With snug fitting lid
- Good chemical resistance
Universal multipurpose bucket without spout.
Frequently Bought Together