Sample container, with lid
15 ml, 20 mm, 80 mm
N° de catalogue 391-3792
Fournisseur: SEMADENI
Aller à
Product Details & Documents
Transparent container made of PS, with attachable blue lid and small spoon made of PE.
- Good chemical resistance
- Good temperature resistance
Spécifications
- Ø ext.:20 mm
- Capacité:15 ml
- Hauteur:80 mm