Waste Bin, PP
Waste bin with handles, gris clair, PP, 60 L, 555×280×585 mm
N° de catalogue SEMA1331
Fournisseur: SEMADENI
Aller à
Product Details & Documents
Bins, made of PP.
- Tough and dimensionally stable
- Smooth surface facilitates easy cleaning
- Stackable conical containers
These versatile and universally applicable plastic containers are suitable for sorting and storage of solid waste.
Spécifications
- Capacité:60 L
- Couleur:gris clair
- Désignation:Waste bin with handles
- Matière:PP
- l×p×h:555×280×585 mm