Anion exchange HPLC columns, Dionex™, IonPac™ AS11-HC
Supplier: DIONEX
Certificates
About this item
AS11-HC columns are used to resolve a larger number of inorganic anions and organic acid anions in complex matrices, sample matrices such as chemical wastewater effluents and fermentation broth solutions contain a variety of inorganic anions and organic acids.
- Requires long retention time with higher resolution
- Ideal for organic acids and anions
- Can be used for monovalent and divalent organic acids
Specifications
- Catalog No:
- DION052961
- DION052962
- DION52963
- DION052960
- Pression max.:
- 4000 psi
- 4000 psi
- 4000 psi
- 4000 psi
- Plage de pH:
- 0 - 14
- 0 - 14
- 0 - 14
- 0 - 14