ContactUsLinkComponent
Protective barrier creams, Sineprint®
  115-1755
undefined
Protective barrier creams, Sineprint®
  115-1755
  • Désignation:
    Protective barrier cream, Sineprint®, 100 ml Tube
  • Cdt:
    12
White, non-greasy barrier cream (oil/water emulsion) for use in workplaces where greasy residue must be avoided, such as in the production of electronics, final inspections, paint shops, or in the paper industry.
  • No greasy residue and absorbs quickly
  • Silicone-free, scented
  • Neutral pH value, contains no mineral oil
 : *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904