Pipette rack, Rotilabo®
Catalog # ROTHCX89.1
Supplier: Roth Carl
Specifications
- Pour:Graduated pipettes
- Couleur:bleu
- l×p×h:291×88×401 mm
- Cdt:1
Specifications
About this item
Pipette rack made of blue coloured ABS.
- For holding graduated pipettes from 0,1 to 25 ml
- With four levels
- With additional holders on both long sides for manual pipetting aids and pipette fillers