Dispenser with top loading hinged lid
Dispenser, PETG, incolore, 305×280×305 mm
Catalog # AKAAAK-260
Supplier: AK
Jump to:
Product Details & Documents
Clear plastic dispenser with top loading hinged lid for use in food and medical areas.
- Clear plastic construction, square design
- Versatile and high impact model
- Wall mount or free standing
Certifications: Approved by FDA and NSF.
Specifications
- Couleur:incolore
- Désignation:Dispenser, PETG
- l×p×h:305×280×305 mm
- Cat. No.:AKAAAK-260
- Type de distributeur:Modèle sur pied / pour montage mural
- Matière:PETG
- PPE type:Multi-use