Bandages, DermaCare®
118-1245
: SOEHNGEN
- Désignation:Bandage pack, medium, DIN, contains bandage (4 m) with wound covering (80×100 mm)
- L×l:4 m × 80 mm
- Cdt:1
Pathogen-free wound covering made of non woven viscose fabric with a high absorbency of 2000 g/m² and hydrophilic PP anti-adhesive non woven fabric on both sides.
- Smooth surface, causes no hypersensitivity reactions
- Physiologically safe, free of chemical binding materials
- EO-sterilised under constant monitoring
- Packed in peel-open packing labelled with an expiry date of 20 years (from the date of manufacture)