Culture bottles with screw cap
Catalog # 215-1913
Supplier: schuett-biotec GmbH
Specifications
- Capacité:100 ml
- Seq:01
- Cdt:10
Specifications
About this item
Clear glass.
- Good chemical resistance
- Good temperature resistance
Complete with aluminium screw cap (autoclavable/sterilisable) or plastic screw cap (steam-autoclavable with 121 °C/sterilisable) and rubber gasket.