Vehicle first aid kits
118-1212
: SOEHNGEN
- Désignation:Vehicle first aid case, ABS, black, DIN 13164
- Dimensions:260×80×170 mm
- Cdt:1
- Poids:0,7 kg
This first aid case is made of temperature-resistant plastic. Sterile contents are marked with an expiration date of 20 years (from the date of manufacture).
: DIN 13164